This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03584470_m1
See other LOC9266585 GE Assays ›
Gene Symbol:
LOC9266585
Gene Name:
histone-lysine N-methyltransferase ASHR1
Gene Aliases:
OSNPB_030704200
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03584470_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC9266585
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015776823.1 XP_015632309.1 5 - 6 733 117
XM_015776824.1 XP_015632310.1 5 - 6 733 117
XM_015776825.1 XP_015632311.1 5 - 6 733 117
GenBank mRNA AK071176.1 - 5 - 6 753 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0