Assay Details
Gene Symbol: LOC4329751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768992.2 | XP_015624478.1 | 18 - 19 | 2365 | 92 | |
GenBank mRNA | AK058407.1 | - | 12 - 13 | 1021 | 92 | |
AK099974.1 | - | 18 - 19 | 2201 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
