Assay Details
Gene Symbol: LOC4339466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783834.2 | XP_015639320.1 | 4 - 5 | 647 | 106 | |
GenBank mRNA | AK070127.1 | - | 4 - 5 | 613 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
