Assay Details
Gene Symbol: LOC4338067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782574.2 | XP_015638060.1 | 3 - 4 | 570 | 67 | |
GenBank mRNA | AB232157.1 | - | 2 - 3 | 415 | 67 | |
AK111723.1 | - | 2 - 3 | 415 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
