This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03561375_g1
See other LOC4339877 GE Assays ›
Gene Symbol:
LOC4339877 LOC4331752
Gene Name:
phosphoglycerate mutase-like protein AT74H
uncharacterized LOC4331752
Gene Aliases:
OSNPB_060109000
OSNPB_030168200
Chromosome Location:
-
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
-
Assay ID Os03561375_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4339877
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK318560.1 - 1743 84
Gene Symbol: LOC4331752
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015772500.2 XP_015627986.1 4 - 5 812 84
GenBank mRNA AK099530.1 - 4 - 5 796 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header