Assay Details
Gene Symbol: LOC4339877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK318560.1 | - | 1970 | 133 |
Gene Symbol: LOC4331752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772500.2 | XP_015627986.1 | 2 - 3 | 585 | 133 | |
GenBank mRNA | AK099530.1 | - | 2 - 3 | 569 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
