Assay Details
Gene Symbol: LOC4330984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767358.2 | XP_015622844.1 | 2 - 3 | 607 | 86 | |
GenBank mRNA | AK105863.1 | - | 2 - 3 | 587 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
