Assay Details
Gene Symbol: LOC4328443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769351.2 | XP_015624837.1 | 6 - 7 | 664 | 152 | |
XM_015769352.2 | XP_015624838.1 | 6 - 7 | 664 | 152 | ||
GenBank mRNA | AK109522.1 | - | 6 - 7 | 610 | 152 | |
DQ004266.1 | - | 6 - 7 | 598 | 152 | ||
JX025003.1 | - | 6 - 7 | 610 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
