This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03559234_m1
See other LOC4328443 GE Assays ›
Gene Symbol:
LOC4328443
Gene Name:
MADS-box transcription factor 29-like
Gene Aliases:
FST, MADS29, OJ1116_A06.26, OSNPB_020170300, OsJ_05554, OsMADS29
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03559234_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4328443
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015769351.2 XP_015624837.1 4 - 5 587 118
XM_015769352.2 XP_015624838.1 4 - 5 587 118
GenBank mRNA AK109522.1 - 4 - 5 533 118
DQ004266.1 - 4 - 5 521 118
JX025003.1 - 4 - 5 533 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0