Assay Details
Gene Symbol: LOC4328443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769351.2 | XP_015624837.1 | 1 - 2 | 78 | 106 | |
XM_015769352.2 | XP_015624838.1 | 1 - 2 | 78 | 106 | ||
GenBank mRNA | AK109522.1 | - | 1 - 2 | 24 | 106 | |
JX025003.1 | - | 1 - 2 | 24 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769351.2, XM_015769352.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
