This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03557442_g1
See other LOC4347510 GE Assays ›
Gene Symbol:
LOC4347510
Gene Name:
nucleotide pyrophosphatase/phosphodiesterase
Gene Aliases:
OSNPB_090506000
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03557442_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4347510
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015795754.2 XP_015651240.1 8 - 9 1217 71
GenBank mRNA AK073281.1 - 8 - 9 982 71
AK073291.1 - 7 - 8 729 71
AK102346.1 - 8 - 9 1191 71
AK106533.1 - 8 - 9 1217 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0