This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03547796_m1
See other LOC4326594 GE Assays ›
Gene Symbol:
LOC4326594
Gene Name:
glucose-1-phosphate adenylyltransferase large subunit 2, cytosolic-like
Gene Aliases:
AGPase, AGPiso, OSNPB_010633100, P0663E10.9
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03547796_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4326594
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015794875.2 XP_015650361.1 3 - 4 1598 139
XM_015794884.2 XP_015650370.1 4 - 5 1948 139
XM_015794892.2 XP_015650378.1 3 - 4 725 139
XM_015794901.2 XP_015650387.1 4 - 5 1954 139
XM_015794909.2 XP_015650395.1 3 - 4 1745 139
XM_015794917.2 XP_015650403.1 3 - 4 1739 139
GenBank mRNA AK071497.1 - 4 - 5 898 139
AY028314.1 - 2 - 3 383 139
EU267956.1 - 2 - 3 381 139

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

nucleotidyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header