Assay Details
Gene Symbol: LOC4326594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794875.2 | XP_015650361.1 | 12 - 13 | 2441 | 69 | |
XM_015794884.2 | XP_015650370.1 | 13 - 14 | 2791 | 69 | ||
XM_015794892.2 | XP_015650378.1 | 12 - 13 | 1568 | 69 | ||
XM_015794901.2 | XP_015650387.1 | 13 - 14 | 2797 | 69 | ||
XM_015794909.2 | XP_015650395.1 | 12 - 13 | 2588 | 69 | ||
XM_015794917.2 | XP_015650403.1 | 12 - 13 | 2582 | 69 | ||
GenBank mRNA | AK071497.1 | - | 13 - 14 | 1741 | 69 | |
AY028314.1 | - | 11 - 12 | 1226 | 69 | ||
EU267956.1 | - | 11 - 12 | 1224 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
