Assay Details
Gene Symbol: LOC4326594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794875.2 | XP_015650361.1 | 10 - 11 | 2279 | 126 | |
XM_015794884.2 | XP_015650370.1 | 11 - 12 | 2629 | 126 | ||
XM_015794892.2 | XP_015650378.1 | 10 - 11 | 1406 | 126 | ||
XM_015794901.2 | XP_015650387.1 | 11 - 12 | 2635 | 126 | ||
XM_015794909.2 | XP_015650395.1 | 10 - 11 | 2426 | 126 | ||
XM_015794917.2 | XP_015650403.1 | 10 - 11 | 2420 | 126 | ||
GenBank mRNA | AK071497.1 | - | 11 - 12 | 1579 | 126 | |
AY028314.1 | - | 9 - 10 | 1064 | 126 | ||
EU267956.1 | - | 9 - 10 | 1062 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
