Assay Details
Gene Symbol: LOC4326594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794875.2 | XP_015650361.1 | 9 - 10 | 2204 | 149 | |
XM_015794884.2 | XP_015650370.1 | 10 - 11 | 2554 | 149 | ||
XM_015794892.2 | XP_015650378.1 | 9 - 10 | 1331 | 149 | ||
XM_015794901.2 | XP_015650387.1 | 10 - 11 | 2560 | 149 | ||
XM_015794909.2 | XP_015650395.1 | 9 - 10 | 2351 | 149 | ||
XM_015794917.2 | XP_015650403.1 | 9 - 10 | 2345 | 149 | ||
GenBank mRNA | AK071497.1 | - | 10 - 11 | 1504 | 149 | |
AY028314.1 | - | 8 - 9 | 989 | 149 | ||
EU267956.1 | - | 8 - 9 | 987 | 149 | ||
U66041.1 | - | 8 - 9 | 968 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
