Assay Details
Gene Symbol: LOC4326030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791944.2 | XP_015647430.1 | 8 - 9 | 1207 | 81 | |
XM_015791954.2 | XP_015647440.1 | 7 - 8 | 1126 | 81 | ||
GenBank mRNA | AK065466.1 | - | 8 - 9 | 1100 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
