Assay Details
Gene Symbol: LOC4326030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791944.2 | XP_015647430.1 | 1 - 2 | 525 | 176 | |
XM_015791954.2 | XP_015647440.1 | 1 - 2 | 525 | 176 | ||
GenBank mRNA | AK065466.1 | - | 2 - 3 | 499 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() |
