Assay Details
Gene Symbol: LOC4326030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791944.2 | XP_015647430.1 | 13 - 14 | 2010 | 87 | |
XM_015791954.2 | XP_015647440.1 | 12 - 13 | 1929 | 87 | ||
GenBank mRNA | AK065466.1 | - | 13 - 14 | 1903 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
