Assay Details
Gene Symbol: LOC4326732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766191.2 | XP_015621677.1 | 8 - 9 | 1035 | 74 | |
XM_015766192.2 | XP_015621678.1 | 8 - 9 | 1034 | 74 | ||
GenBank mRNA | AK064461.1 | - | 3 - 4 | 909 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
