Assay Details
Gene Symbol: LOC4337878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785288.2 | XP_015640774.1 | 1 - 2 | 539 | 61 | |
GenBank mRNA | AK067837.1 | - | 1 - 2 | 540 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
