Assay Details
Gene Symbol: LOC4334871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775317.2 | XP_015630803.1 | 6 - 7 | 958 | 83 | |
GenBank mRNA | AK100706.1 | - | 6 - 7 | 781 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
