This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03540902_m1
See other LOC4331459 GE Assays ›
Gene Symbol:
LOC4331459 LOC4337686
Gene Name:
50S ribosomal protein L5, chloroplastic-like
phosphoinositide phospholipase C 2
Gene Aliases:
OSNPB_030125000, rpl5
OSNPB_050127200
Chromosome Location:
-
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
-
Assay ID Os03540902_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4331459
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015772370.2 XP_015627856.1 2 - 3 574 81
GenBank mRNA AF095708.1 - 2 - 3 508 81
AK059557.1 - 2 - 3 516 81
AK070779.1 - 2 - 3 541 81
AK098999.1 - 2 - 3 519 81
AK104137.1 - 2 - 3 512 81
Gene Symbol: LOC4337686
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK072141.1 - 595 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0