This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03540226_g1
See other LOC4330164 GE Assays ›
Gene Symbol:
LOC4330164
Gene Name:
glutamate dehydrogenase 3, mitochondrial-like
Gene Aliases:
GDH3, OSNPB_020650900
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03540226_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4330164
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015767290.1 XP_015622776.1 7 - 8 1180 95
GenBank mRNA AB035927.1 - 7 - 8 1103 95
AK103028.1 - 7 - 8 1077 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0