Assay Details
Gene Symbol: LOC4336156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015779498.2 | XP_015634984.1 | 6 - 7 | 2490 | 93 | |
GenBank mRNA | AK105466.1 | - | 5 - 6 | 1360 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
