Assay Details
Gene Symbol: LOC4336057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780820.2 | XP_015636306.1 | 4 - 5 | 651 | 70 | |
GenBank mRNA | AK243669.1 | - | 3 - 4 | 1342 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
