Assay Details
Gene Symbol: LOC4330014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767241.2 | XP_015622727.1 | 6 - 7 | 1058 | 72 | |
GenBank mRNA | AK120798.1 | - | 6 - 7 | 998 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
