Assay Details
Gene Symbol: LOC4326615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766117.2 | XP_015621603.1 | 1 - 2 | 314 | 59 | |
GenBank mRNA | AK067476.1 | - | 1 - 2 | 313 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
