This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03524569_m1
See other LOC4351908 GE Assays ›
Gene Symbol:
LOC4351908
Gene Name:
serine carboxypeptidase 1-like
Gene Aliases:
CBP1, OSNPB_120257000
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03524569_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4351908
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015762694.2 XP_015618180.1 9 - 10 1224 70
XM_015762695.2 XP_015618181.1 9 - 10 1219 70
GenBank mRNA AK064874.1 - 9 - 10 1142 70
AK072798.1 - 6 - 7 670 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0