Assay Details
Gene Symbol: LOC4347035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795677.2 | XP_015651163.1 | 1 - 2 | 256 | 54 | |
GenBank mRNA | AK058290.1 | - | 1 - 2 | 213 | 54 | |
AK104919.1 | - | 1 - 2 | 183 | 54 | ||
AK121091.1 | - | 1 - 2 | 250 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
