Assay Details
Gene Symbol: LOC9269631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757447.2 | XP_015612933.1 | 5 - 6 | 1970 | 67 | |
XM_015757448.2 | XP_015612934.1 | 5 - 6 | 1971 | 67 | ||
XM_015757450.2 | XP_015612936.1 | 4 - 5 | 1691 | 67 | ||
GenBank mRNA | AK060203.1 | - | 3 - 4 | 1082 | 67 | |
AK100608.1 | - | 4 - 5 | 2379 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
