Assay Details
Gene Symbol: LOC112936009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK072553.1 | - | 1 - 2 | 358 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: LOC112936009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK072553.1 | - | 1 - 2 | 358 | 130 |
Set Membership: |
![]() ![]() ![]() ![]() |