Assay Details
Gene Symbol: LOC4331872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775124.1 | XP_015630610.1 | 3 - 4 | 698 | 86 | |
XM_015775125.1 | XP_015630611.1 | 3 - 4 | 602 | 86 | ||
GenBank mRNA | AJ293816.1 | - | 3 - 4 | 351 | 86 | |
AJ536158.1 | - | 3 - 4 | 384 | 86 | ||
AK242211.1 | - | 3 - 4 | 603 | 86 | ||
AY345221.1 | - | 3 - 4 | 384 | 86 | ||
AY345222.1 | - | 3 - 4 | 384 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
