This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03511091_m1
See other LOC4331872 GE Assays ›
Gene Symbol:
LOC4331872
Gene Name:
MADS-box transcription factor 47-like
Gene Aliases:
OSNPB_030186600, OsMADS47, mads47, mdp
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03511091_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4331872
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015775124.1 XP_015630610.1 1 - 2 553 63
XM_015775125.1 XP_015630611.1 1 - 2 457 63
GenBank mRNA AJ293816.1 - 1 - 2 206 63
AJ536158.1 - 1 - 2 239 63
AK242211.1 - 1 - 2 458 63
AY345221.1 - 1 - 2 239 63
AY345222.1 - 1 - 2 239 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0