Assay Details
Gene Symbol: LOC4330061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770436.2 | XP_015625922.1 | 12 - 13 | 1135 | 98 | |
GenBank mRNA | AK101701.1 | - | 12 - 13 | 1142 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
