Assay Details
Gene Symbol: LOC9269678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764229.2 | XP_015619715.1 | 9 - 10 | 4633 | 80 | |
XM_015764230.2 | XP_015619716.1 | 7 - 8 | 4295 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
