Assay Details
Gene Symbol: LOC9269678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764229.2 | XP_015619715.1 | 8 - 9 | 3412 | 106 | |
XM_015764230.2 | XP_015619716.1 | 6 - 7 | 3074 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
