Assay Details
Gene Symbol: LOC4328747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771617.2 | XP_015627103.1 | 1 - 2 | 324 | 125 | |
GenBank mRNA | AK072526.1 | - | 1 - 2 | 328 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
