Assay Details
Gene Symbol: LOC4335698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778772.2 | XP_015634258.1 | 4 - 5 | 505 | 136 | |
GenBank mRNA | AK107750.1 | - | 4 - 5 | 505 | 136 | |
AK240828.1 | - | 537 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
