This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03492813_g1
See other LOC4335556 GE Assays ›
Gene Symbol:
LOC4335556
Gene Name:
70 kDa peptidyl-prolyl isomerase
Gene Aliases:
OSNPB_040352400
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03492813_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4335556
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015778615.2 XP_015634101.1 8 - 9 1469 65
GenBank mRNA AF140495.2 - 8 - 9 1396 65
AK100844.1 - 8 - 9 1399 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0