Assay Details
Gene Symbol: LOC4331214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771005.2 | XP_015626491.1 | 9 - 10 | 2284 | 93 | |
GenBank mRNA | AK101090.1 | - | 9 - 10 | 2270 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
