Assay Details
Gene Symbol: LOC4339897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788367.2 | XP_015643853.1 | 2 - 3 | 526 | 87 | |
GenBank mRNA | AK061381.1 | - | 2 - 3 | 367 | 87 | |
AK069439.1 | - | 2 - 3 | 357 | 87 | ||
AK103951.1 | - | 2 - 3 | 342 | 87 | ||
AK104422.1 | - | 2 - 3 | 354 | 87 | ||
AK104612.1 | - | 2 - 3 | 332 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
