This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03477592_m1
See other LOC4331005 GE Assays ›
Gene Symbol:
LOC4331005
Gene Name:
DEAD-box ATP-dependent RNA helicase 1-like
Gene Aliases:
OJ1004_E04.4, OJ1695_H09.19, OSNPB_020795900
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03477592_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4331005
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015771853.2 XP_015627339.1 7 - 8 1506 104
XM_015771854.2 XP_015627340.1 7 - 8 1506 104
XM_015771856.2 XP_015627342.1 7 - 8 1506 104
GenBank mRNA AK105305.1 - 7 - 8 1497 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0