This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03476651_m1
See other LOC4352218 GE Assays ›
Gene Symbol:
LOC4352218
Gene Name:
DEAD-box ATP-dependent RNA helicase 28-like
Gene Aliases:
OSNPB_120481100, OsJ_36091
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03476651_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4352218
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015764854.2 XP_015620340.1 5 - 6 1019 116
GenBank mRNA AK073151.1 - 5 - 6 1007 116
AK099385.1 - 5 - 6 1010 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header