Assay Details
Gene Symbol: LOC4327645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765933.2 | XP_015621419.1 | 6 - 7 | 956 | 69 | |
GenBank mRNA | AK100326.1 | - | 6 - 7 | 956 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
