Assay Details
Gene Symbol: LOC4327645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765933.2 | XP_015621419.1 | 13 - 14 | 2353 | 75 | |
XM_015765935.2 | XP_015621421.1 | 13 - 14 | 2200 | 75 | ||
GenBank mRNA | AK100326.1 | - | 13 - 14 | 2353 | 75 | |
GQ267545.1 | - | 12 - 13 | 1985 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
