Assay Details
Gene Symbol: LOC4327645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765933.2 | XP_015621419.1 | 10 - 11 | 2200 | 66 | |
XM_015765935.2 | XP_015621421.1 | 10 - 11 | 2047 | 66 | ||
XM_015765936.2 | XP_015621422.1 | 10 - 11 | 2047 | 66 | ||
GenBank mRNA | AK100326.1 | - | 10 - 11 | 2200 | 66 | |
GQ267545.1 | - | 9 - 10 | 1832 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
