Assay Details
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271482.2 | XP_008269704.2 | 3 | 1142 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271482.2 | XP_008269704.2 | 3 | 1142 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |