This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06792200_m1
See other CAB39L GE Assays ›
Gene Symbol:
CAB39L
Gene Name:
calcium binding protein 39 like
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 2983351 - 3105084 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06792200_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAB39L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002720727.3 XP_002720773.1 1180 65
XM_008273843.2 XP_008272065.1 917 65
XM_008273844.2 XP_008272066.1 1016 65
XM_008273846.2 XP_008272068.1 726 65
XM_017350279.1 XP_017205768.1 920 65
XM_017350280.1 XP_017205769.1 954 65
XM_017350281.1 XP_017205770.1 822 65
XM_017350282.1 XP_017205771.1 819 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0