Assay Details
Gene Symbol: SRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262939.2 | XP_008261161.1 | 7 - 8 | 1814 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: SRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262939.2 | XP_008261161.1 | 7 - 8 | 1814 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |