This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06781689_g1
See other TMEM200B GE Assays ›
Gene Symbol:
TMEM200B
Gene Name:
transmembrane protein 200B
Gene Aliases:
-
Chromosome Location:
Chr.13: 136456262 - 136460294 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06781689_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM200B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008265859.2 XP_008264081.1 2 - 3 1016 80
XM_008265860.2 XP_008264082.1 2 - 3 757 80
XM_008265861.2 XP_008264083.1 2 - 3 152 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008265859.2, XM_008265860.2, XM_008265861.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0